missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Chemicon™ QCM ECMatrix Cell Invasion Assay, 24-well (8μm), Colorimetric
For evaluating the invasion of tumor cells through a basement membrane model
Supplier: MilliporeSigma™ ECM550
Specifications
2°C to 8°C | |
Colorimetric | |
Forceps, Invasion Chamber, Cell Stain, Cotton Swabs |
QCM ECMatrix Cell Invasion Assay | |
For evaluating the invasion of tumor cells through a basement membrane model | |
1/Pk. |