MilliporeSigma™ Chemicon™ Human Corneal Epithelial Cells
provides an ideal serum-free culture model without antimicrobials or phenol red when used with the EpiGRO Ocular Epithelia Complete Media Kit.
Supplier: MilliporeSigma™ SCCE016
Description
Milliporeâs Human Corneal Epithelial Cell Line provides an ideal serum-free culture model without antimicrobials or phenol red when used with the EpiGRO™ Ocular Epithelia Complete Media Kit. These cells are perfect for the accurate studies of cell-matrix interactions, gene regulation and tissue development, drug development, and validation of alternative methods in toxicology.Specifications
Cryopreserved | |
Cell Culture | |
500,000 cells | |
Human Corneal Epithelial Cells |