missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Calbiochem™ DNP-Albumin Conjugate, Bovine

For production of antibodies specific for DNP and albumin

Supplier:  MilliporeSigma™ 324101100MG

Catalog No. 32-410-1100MG



Description

Description

  • Contains >30 DNP-groups per molecule protein
  • Form: Dry amber solid
  • Soluble in water
Specifications

Specifications

DNP-Albumin Conjugate, Bovine
100 mg
2°C to 8°C
SDS
Documents

Documents

Promotions

Promotions