missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore™ Amicon™ Ultra-15 Centrifugal Filter Units

Designed for protein purification, protein concentration and desalting. EMD Millipore Amicon™ Ultra-15 Centrifugal Filter Units are constructed with Ultracel membranes, styrene/butadiene housing and polypropylene tubes and polyethylene caps and liners. Available in many MWCO levels.

$121.00 - $1,230.00

Specifications

Membrane Material Ultracel-PL Regenerated Cellulose
For Use With (Application) Protein purification, protein concentration and desalting
Housing Material
  • Housing: Styrene/butadiene
  • Tube: Polypropylene
  • Cap and liner: Polyethylene
Includes Assembled with centrifuge tubes and caps
Surface Area 7.6cm2
View More Specs

Includes: Assembled with centrifuge tubes and caps

Products 15 Description

Description

  • High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
  • Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
  • Heat-sealed membrane eliminates possibility of downstream extractables
  • Universal rotor compatibility
  • High retentate recovery of >90%
  • High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200µL)
  • Easy sample monitoring due to translucent housing and volume gradations
  • Direct pipetter sample access eliminates processing step to recover concentrate
Centrifugal filter enables
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates
  • Concentration of dilute or prepurified proteins from column eluents
  • Desalting and buffer exchange
Specifications

Specifications

Ultracel-PL Regenerated Cellulose
  • Housing: Styrene/butadiene
  • Tube: Polypropylene
  • Cap and liner: Polyethylene
7.6cm2
121mm
Protein purification, protein concentration and desalting
Assembled with centrifuge tubes and caps
29.7mm
15mL