EMD Millipore™ Amicon™ Ultra-0.5 Centrifugal Filter Units
Designed for protein purification, protein concentration and desalting. EMD Millipore Amicon™ Ultra-0.5 Centrifugal Filter Units feature reverse spin capability and are available at two MWCO levels. Constructed with Ultracel membranes, styrene/butadiene housing and polypropylene caps and tubes.
$55.00 - $1,900.00
Specifications
Membrane Material Regenerated Cellulose
For Use With (Application) Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms; Purification of macromolecular components found in tissue culture extracts or cell lysates, removal of primer, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC: Desalting and buffer exchange
Includes Two microcentrifuge tubes
Length (English) 0.196 in.
Length (Metric) 4.99cm
View More Specs
Includes: Two microcentrifuge tubes
Products 20
Catalog Number Mfr. No. MWCO Price Quantity  
Catalog Number Mfr. No. MWCO Price Quantity  
UFC501008 View Documents EMD Millipore™
UFC501008
10,000 d Pack of 8 Each for $56.00
UFC5010BK View Documents EMD Millipore™
UFC5010BK
10,000 d Pack of 500 Each for $1,900.00
UFC500324 View Documents EMD Millipore™
UFC500324
3,000 d Pack of 24 Each for $146.00
UFC500396 View Documents EMD Millipore™
UFC500396
3,000 d Pack of 96 Each for $447.00
UFC5003BK View Documents EMD Millipore™
UFC5003BK
3,000 d Pack of 500 Each for $1,900.00
UFC501024 View Documents EMD Millipore™
UFC501024
10,000 d Pack of 24 Each for $146.00
UFC500308 View Documents EMD Millipore™
UFC500308
3,000 d Pack of 8 Each for $56.00
UFC501096 View Documents EMD Millipore™
UFC501096
10,000 d Pack of 96 Each for $474.00
UFC503024 View Documents EMD Millipore™
UFC503024
30,000 d Pack of 24 Each for $144.00
UFC503008 View Documents EMD Millipore™
UFC503008
30,000 d Pack of 8 Each for $55.00
UFC503096 View Documents EMD Millipore™
UFC503096
30,000 d Pack of 96 Each for $450.01
UFC5030BK View Documents EMD Millipore™
UFC5030BK
30,000 d Pack of 500 Each for $1,900.00
UFC505008 View Documents EMD Millipore™
UFC505008
50,000 d Pack of 8 Each for $55.00
UFC505024 View Documents EMD Millipore™
UFC505024
50,000 d Pack of 24 Each for $144.00
UFC505096 View Documents EMD Millipore™
UFC505096
50,000 d Pack of 96 Each for $447.00
UFC5050BK View Documents EMD Millipore™
UFC5050BK
50,000 d Pack of 500 Each for $1,900.00
UFC510008 View Documents EMD Millipore™
UFC510008
100,000 d Pack of 8 Each for $55.00
UFC510024 View Documents EMD Millipore™
UFC510024
100,000 d Pack of 24 Each for $144.00
UFC5100BK View Documents EMD Millipore™
UFC5100BK
100,000 d Pack of 500 Each for $1,900.00
UFC510096 View Documents EMD Millipore™
UFC510096
100,000 d Pack of 96 Each for $471.00
Description
Description
  • Concentrates 500μL down to 15μL
  • Concentration factor 25x to 30x
  • Fixed angle rotor: 14,000 × g for concentration spin; 1000 × g for reverse spin
  • High sample recovery (greater than 90% of dilute starting solution)
  • Fast processing time - 10 to 30 minutes
  • Reverse spin capability provides consistent recoveries
  • Enables the concentration of proteins with a membrane molecular weight limit of 3 kDa or 10 kDa
Applications:
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates, removal of primer, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC
  • Desalting and buffer exchange
Specifications
Specifications
Regenerated Cellulose
Two microcentrifuge tubes
4.99cm
1cm2
Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms; Purification of macromolecular components found in tissue culture extracts or cell lysates, removal of primer, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC: Desalting and buffer exchange
0.196 in.
14,000xG for concentration spin; 1000xG for reverse spin