missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco Clamp Retort Combined

Diecast alloy clamp retort, PVC coated jaws with boss head which accommodates rods up to 15mm in diameter.

Manufacturer:  Eisco CH0688

Catalog No. S02629



Description

Description

  • Diecast alloy
  • Rubber-lined jaws with boss head
  • Accomodates rods up to 15mm in diameter
Specifications

Specifications

11.81 in.
30cm
Diecasted Alloy, PVC, Rubber
Clamp Retort Combined
Documents