Edvotek™ Edvotek™ MV10 Protein Electrophoresis Apparatus
Seamless vertical electrophoresis unit is designed for separation of proteins on a single polyacrylamide gel.
Supplier:  Edvotek™ 581
Catalog No. S90257ND
Includes:
  • Seamless injection-molded base
  • Safety interlock cover
  • Corrosion-resistant, all-platinum electrodes with safety-insulated leads
Description
Description
Re-designed MV10 vertical protein electrophoresis unit capable of holding one vertical polyacrylamide gel, the unit has color coded parts to ensure proper orientation.
  • Gel support cassette clip - holds gel securely, allows for easy placement and removal of gel
  • New, sleek design
  • Domed lid with sturdy overmold grip feature
  • Modified shapes improves gel running speed by 10-15%
  • Stabilizing feet - raises unit off bench to promote cooling and improves stabilization
  • Placement of color-coded anode/cathode sides completely eliminates the chance of improper or reverse lid placement
  • Electrode lead 90° right angle shape improves stability by lowering the overall profile and thus minimizes tipping
  • Patent pending
Warranty
Manufacturer's limited lifetime warranty and free technical support.
Specifications
Specifications
Industry Standard 9 x 10 cm PreCast Gel Cassettes (10 samples)
Seamless injection-molded base, safety interlock cover, corrosion resistant, all-Platinum electrodes with safety-insulated Leads
Lifetime