missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
EDTA Titrant, 0.05M solution, Macron Fine Chemicals™
macron
$387.89 - $387.89
Chemical Identifiers
CAS | 60-00-4 |
---|---|
Molecular Formula | C10H16N2O8 |
Molecular Weight (g/mol) | 292.24 |
MDL Number | MFCD00003541 |
InChI Key | KCXVZYZYPLLWCC-UHFFFAOYSA-N |
Synonym | edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol |
PubChem CID | 6049 |
ChEBI | CHEBI:42191 |
IUPAC Name | 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid |
SMILES | OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
MKH1151
|
Avantor Macron
H11505 |
1 L |
Each for $387.89
|
|
|||||
Chemical Identifiers
60-00-4 | |
292.24 | |
KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
6049 | |
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid |
C10H16N2O8 | |
MFCD00003541 | |
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol | |
CHEBI:42191 | |
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Specifications
Colorless | |
C10H16N2O8 | |
860mg/mL | |
KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid | |
6049 | |
292.24 | |
Crystals |
60-00-4 | |
MFCD00003541 | |
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol | |
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O | |
292.24 | |
CHEBI:42191 | |
1 L | |
EDTA Titrant, 0.01M solution |