missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ EASY-Spray™ Transfer Lines

Tune Thermo Scientific™ EASY-Spray™ source-equipped mass spectrometers more easily using Thermo Scientific™ EASY-Spray™ transfer lines.

Manufacturer:  Thermo Scientific™ ES792

 View more versions of this product

Catalog No. ES792


 

Description

Description

Tune Thermo Scientific™ EASY-Spray™ source-equipped mass spectrometers more easily using Thermo Scientific™ EASY-Spray™ transfer lines. The transfer lines act as a no-separation sprayer, facilitating introduction of external tuning compounds while eliminating troublesome handling and alignment of traditional needles.

  • Available in nanoflow and microflow formats to meet a wider range of tuning requirements
  • Thermo Scientific™ Dionex™ nanoViper™ fitting makes installation easy
  • Precision positioned glass emitter for excellent spray performance
  • Integrated zero-dead-volume unions ensure reliability
Specifications

Specifications

-
-
N/A
-
500mm
Emitter
-
0.075mm
N/A
-
-
-
N/A
Unpacked
EASY-Spray Emitter
Documents

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Specifications, terms, and pricing are subject to change. Not all products are available in all countries. Please consult your local sales representative for details.