missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ DYNAL™ DynaMag™ DynaMag - Spin Magnet DFS Item

Optimized for efficient magnetic separation of all types of Dynabeads (range 1–4.5μm in diameter) in small sample volumes (1.5mL) in microcentrifuge tubes

Manufacturer:  Invitrogen™ LS12320D

Catalog No. 12320D


 

Includes: One DynaMag-Spin magnet

Description

Description

DynaMag-Spin magnet is optimized for efficient magnetic separation of all types of Dynabeads (range 1–4.5μm in diameter) in small sample volumes (1.5mL) in microcentrifuge tubes. The DynaMag-Spin magnet combines a strong magnetic attraction for efficient separations with a flexible and smart ergonomic design. The magnet fits comfortably in the palm of your hand, and the angle of the magnetic field ensures efficient isolation of proteins and nucleic acids in small sample volumes.

  • Optimal working volume: 10–1,500 μL
  • Holds up to six standard 1.5mL microcentrifuge tubes in numbered spaces
  • Good control and visibility of samples
  • Sample rack can be quickly removed from the magnet in the base for resuspension, vortexing, or manual sample shaking

DNA and RNA Purification and Analysis, DNA Extraction, Sequence-Specific DNA or RNA Purification, Sequencing Reaction Purification

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

DNA and RNA Purification and Analysis, DNA Extraction, Sequence-Specific DNA or RNA Purification, Sequencing Reaction Purification
One DynaMag-Spin magnet
1 each
Store at room temperature
Documents

For Research Use Only. Not for use in diagnostic procedures.