missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ 224 Series Lab Coats
Protect against particulates and other contaminants in controlled environments.
$244.75 - $259.42
Specifications
Type | Lab Coat |
---|---|
Material | Tyvek |
Color | White |
Closure Type | Snaps |
Disposable | Single-use |
Alert:
Should not be worn around open flame, heat source, sparks or ignition sources.
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17988026
|
DuPont™
IC224SWH2X00300B |
2X-Large |
Case of 30 Each for $259.41
|
|
|||||
17988001
|
DuPont™
IC224SWHMD00300B |
Medium |
Case of 30 Each for $259.42
|
|
|||||
17988000
|
DuPont™
IC224SWHSM00300B |
Small |
Case of 30 Each for $244.75
|
|
|||||
17988025
|
DuPont™
IC224SWHXL00300B |
X-Large |
Case of 30 Each for $244.75
|
|
|||||
17988024
|
DuPont™
IC224SWHLG00300B |
Large |
Case of 30 Each for $244.75
|
|
|||||
Description
Light and durable labcoats provide upper body coverage, while staying comfortable.
- Spinning process creates barrier to particles, micro-organisms and nonhazardous liquids
- Lowest level particle shedding
- Serged seams
- Five-snap closure
- Laydown collar
- Two lower front pockets and one chest pocket
- Raglan sleeves
Applications:
Use in controlled environments with high standards for particle and microbiological contamination control; Pharmaceutical, biotech, medical device, electronic settings.
Specifications
Lab Coat | |
White | |
Single-use | |
3 | |
Serged | |
Class 1000 (ISO 6) |
Tyvek | |
Snaps | |
Non-sterile | |
Lay Down | |
1910.120 App B, 1910.1450 App A |