missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ ProShield™ NexGen™ Sleeves
Provide protection from dry particulates and nonhazardous aerosols and liquids.
Supplier: DuPont™ NG500SWH00020000
Description
DuPont ProShield NexGen is liquid resistant and provides a true aerosol barrier, with >99% holdout of particles down to 0.35μm. Proprietary microporous technology gives sleeves effective protective properties at an economical price. Perfect for applications involving nontoxic liquid or spray hazards.
Sleeves
- Elastic at both ends
- Length: 18 in. (46cm)
- White
Specifications
General Purpose Sleeve | |
Single-use | |
10 mil |
Microporous Film Laminate | |
Serged | |
100 Pairs/Cs. |