missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Drierite™, without indicator, 8 mesh, ACROS Organics™

$66.96 - $643.62

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.134
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium;sulfate
SMILES [O-]S(=O)(=O)[O-].[Ca+2]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC219050020 SDS View Documents Acros Organics
219050020
2kg Glass Bottle Each for $171.87
AC219055000 SDS View Documents Acros Organics
219055000
500g Glass bottle Each for $66.96
AC219050100 SDS View Documents Acros Organics
AC219050100
10kg Plastic drum Each for $643.62
Description

Description

Adsorbent, Indicator
Specifications

Chemical Identifiers

7778-18-9
136.134
24497
calcium;sulfate
CaO4S
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
CHEBI:31346
[O-]S(=O)(=O)[O-].[Ca+2]

Specifications

7778-18-9
Typical chemical analysis: CaO: 40.80%, SO3: 58.80%, CO2: 0.60%, R2O: 0.14%, SiO2 and insolubles: 0.15%, NaCl: 0.18%
CaO4S
Solubility in water: ca. 0.25g/L (20°C). Other solubilities: insoluble in organic liquids, slightly soluble in glycerol
[O-]S(=O)(=O)[O-].[Ca+2]
136.134
CHEBI:31346
Granular
1450.0°C
2kg
Glass Bottle
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
calcium;sulfate
24497
136.14
Drierite™
Safety and Handling

Safety and Handling