missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals DL-Proline, 99%
$71.67 - $237.06
Chemical Identifiers
CAS | 609-36-9 |
---|---|
Molecular Formula | C5H9NO2 |
Molecular Weight (g/mol) | 115.132 |
MDL Number | MFCD00005250 |
InChI Key | ONIBWKKTOPOVIA-UHFFFAOYSA-N |
Synonym | dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275 |
PubChem CID | 614 |
ChEBI | CHEBI:26271 |
IUPAC Name | pyrrolidine-2-carboxylic acid |
SMILES | C1CC(NC1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC174030050
|
Thermo Scientific Chemicals
174030050 |
5 g | Glass Bottle |
N/A
|
|
||||
AC174030250
|
Thermo Scientific Chemicals
174030250 |
25 g | Glass Bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
609-36-9 | |
115.132 | |
ONIBWKKTOPOVIA-UHFFFAOYSA-N | |
614 | |
pyrrolidine-2-carboxylic acid |
C5H9NO2 | |
MFCD00005250 | |
dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275 | |
CHEBI:26271 | |
C1CC(NC1)C(=O)O |
Specifications
208.0°C to 210.0°C | |
White | |
99% | |
Authentic | |
22, 4 | |
15, 7895 | |
dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275 | |
ONIBWKKTOPOVIA-UHFFFAOYSA-N | |
pyrrolidine-2-carboxylic acid | |
115.132 | |
CHEBI:26271 | |
99% | |
5 g | |
DL-Proline, 99% |
609-36-9 | |
20ppm max. | |
C5H9NO2 | |
MFCD00005250 | |
09,393 | |
Glass Bottle | |
Solubility in water: soluble in water. Other solubilities: soluble in alcohol, sparingly soluble in chloroform, benzene and, acetone, insoluble in ether | |
C1CC(NC1)C(=O)O | |
0.1% max. | |
614 | |
115.13 | |
0.5% max. (K.F.) | |
Crystalline Powder |
Safety and Handling
EINECSNumber : 210-189-3
TSCA : TSCA
RUO – Research Use Only