missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-beta-Proline ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals
$48.33 - $161.92
Chemical Identifiers
CAS | 80028-44-0 |
---|---|
Molecular Formula | C7H14ClNO2 |
Molecular Weight (g/mol) | 179.644 |
MDL Number | MFCD14585012 |
InChI Key | AYYKCALHTPKNOI-UHFFFAOYSA-N |
Synonym | ethyl pyrrolidine-3-carboxylate hydrochloride, dl-beta-proline ethyl ester hydrochloride, ethyl pyrrolidine-3-carboxylate hcl, 3-pyrrolidinecarboxylic acid, ethyl ester, hydrochloride, pubchem18711, ethylpyrrolidine-3-carboxylate hydrochloride, +/--pyrrolidine-3-carboxylic acid ethyl ester hy, 3-pyrrolidinecarboxylic acid ethyl ester hydrochloride |
PubChem CID | 12896003 |
IUPAC Name | ethyl pyrrolidine-3-carboxylate;hydrochloride |
SMILES | CCOC(=O)C1CCNC1.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH57113MD
|
Thermo Scientific Chemicals
H57113MD |
250 mg |
Each for $48.33
|
|
|||||
AAH5711303
|
Thermo Scientific Chemicals
H5711303 |
1 g |
Each for $161.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
80028-44-0 | |
179.644 | |
AYYKCALHTPKNOI-UHFFFAOYSA-N | |
12896003 | |
CCOC(=O)C1CCNC1.Cl |
C7H14ClNO2 | |
MFCD14585012 | |
ethyl pyrrolidine-3-carboxylate hydrochloride, dl-beta-proline ethyl ester hydrochloride, ethyl pyrrolidine-3-carboxylate hcl, 3-pyrrolidinecarboxylic acid, ethyl ester, hydrochloride, pubchem18711, ethylpyrrolidine-3-carboxylate hydrochloride, +/--pyrrolidine-3-carboxylic acid ethyl ester hy, 3-pyrrolidinecarboxylic acid ethyl ester hydrochloride | |
ethyl pyrrolidine-3-carboxylate;hydrochloride |
Specifications
80028-44-0 | |
MFCD14585012 | |
AYYKCALHTPKNOI-UHFFFAOYSA-N | |
ethyl pyrrolidine-3-carboxylate;hydrochloride | |
12896003 | |
97% | |
DL-beta-Proline ethyl ester hydrochloride |
C7H14ClNO2 | |
ethyl pyrrolidine-3-carboxylate hydrochloride, dl-beta-proline ethyl ester hydrochloride, ethyl pyrrolidine-3-carboxylate hcl, 3-pyrrolidinecarboxylic acid, ethyl ester, hydrochloride, pubchem18711, ethylpyrrolidine-3-carboxylate hydrochloride, +/--pyrrolidine-3-carboxylic acid ethyl ester hy, 3-pyrrolidinecarboxylic acid ethyl ester hydrochloride | |
CCOC(=O)C1CCNC1.Cl | |
179.644 | |
179.65 | |
250 mg |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only