Learn More
Diphenyl phosphate, 97%, Thermo Scientific Chemicals
$70.48 - $634.10
Chemical Identifiers
CAS | 838-85-7 |
---|---|
Molecular Formula | C12H11O4P |
Molecular Weight (g/mol) | 250.19 |
MDL Number | MFCD00003033 |
InChI Key | ASMQGLCHMVWBQR-UHFFFAOYSA-N |
Synonym | diphenyl phosphate, phosphoric acid, diphenyl ester, phenyl hydrogen phosphate, phenyl phosphate pho 2 ho po, diphenoxyphosphinic acid, phosphoric acid diphenyl ester, diphenylphosphoric acid, phosphoric acid diphenyl, dsstox_cid_28182 |
PubChem CID | 13282 |
IUPAC Name | diphenyl hydrogen phosphate |
SMILES | OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0212106
|
Thermo Scientific Chemicals
L0212106 |
5 g |
N/A
|
|
|||||
AAL0212114
|
Thermo Scientific Chemicals
L0212114 |
25 g |
N/A
|
|
|||||
AAL0212122
|
Thermo Scientific Chemicals
L0212122 |
100 g |
N/A
|
|
|||||
Description
Diphenyl phosphate is used in the manufacture of phosphoric acid diesters such as triphenyl phosphate, trixylenyl phosphate, isodecyl diphenyl phosphate, cresyl diphenyl phosphate and isopropylphenyl diphenyl phosphate. It is widely used as a protective agent for hydroxyl group in organic synthesis. It finds application as an additive for paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiphenyl phosphate is used in the manufacture of phosphoric acid diesters such as triphenyl phosphate, trixylenyl phosphate, isodecyl diphenyl phosphate, cresyl diphenyl phosphate and isopropylphenyl diphenyl phosphate. It is widely used as a protective agent for hydroxyl group in organic synthesis. It finds application as an additive for paints and coatings.
Solubility
Soluble in benzene. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
838-85-7 | |
250.19 | |
ASMQGLCHMVWBQR-UHFFFAOYSA-N | |
13282 | |
OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1 |
C12H11O4P | |
MFCD00003033 | |
diphenyl phosphate, phosphoric acid, diphenyl ester, phenyl hydrogen phosphate, phenyl phosphate pho 2 ho po, diphenoxyphosphinic acid, phosphoric acid diphenyl ester, diphenylphosphoric acid, phosphoric acid diphenyl, dsstox_cid_28182 | |
diphenyl hydrogen phosphate |
Specifications
838-85-7 | |
C12H11O4P | |
5 g | |
diphenyl phosphate, phosphoric acid, diphenyl ester, phenyl hydrogen phosphate, phenyl phosphate pho 2 ho po, diphenoxyphosphinic acid, phosphoric acid diphenyl ester, diphenylphosphoric acid, phosphoric acid diphenyl, dsstox_cid_28182 | |
ASMQGLCHMVWBQR-UHFFFAOYSA-N | |
diphenyl hydrogen phosphate | |
13282 | |
97% |
62°C to 70°C | |
MFCD00003033 | |
1379164 | |
Soluble in benzene. Insoluble in water. | |
OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1 | |
250.19 | |
250.19 | |
Diphenyl phosphate |
Safety and Handling
EINECSNumber : 212-657-2
RTECSNumber : TC5470000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only