missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dinonyl phthalate, EMD-Millipore

Manufacturer:  Millipore Sigma 1096690100

Catalog No. M1096690100


 

Specifications

Specifications

Dinonyl phthalate
C26H42O4
DROMNWUQASBTFM-UHFFFAOYSA-N
dinonyl benzene-1,2-dicarboxylate
6787
Glass
Solvent: Acetone/Chloroform
84-76-4
DNP, Phthalic acid dinonyl ester
CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC
418.618
418.61
Density: 0.98g/cm3 (20°C)
100mL

Chemical Identifiers

84-76-4
418.618
DNP, Phthalic acid dinonyl ester
C26H42O4
DROMNWUQASBTFM-UHFFFAOYSA-N
6787
Safety and Handling

Safety and Handling

Recommended Storage : Store without restrictions

Documents