missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyltin dichloride, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 07114204

 View more versions of this product

Catalog No. AA7114204



Description

Description

Used as a pharmaceutical intermediate. It is employed as a coating, plastic and rubber auxilliary agent. Used as methyl Tin stabilizer intermediates, catalysts for esterification reaction, raw material for glass coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dimethyltin dichloride
103°C to 105°C
C2H6Cl2Sn
2 g
3660508
dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity
PKKGKUDPKRTKLJ-UHFFFAOYSA-L
dichloro(dimethyl)stannane
12955
Crystalline
753-73-1
185°C to 190°C
MFCD00000517
UN3146
Moisture sensitive
Slightly soluble in water (20g/L (20°C)). Solubility in other solvents (0.1g/ml in methanol).
C[Sn](C)(Cl)Cl
219.68
219.69

Chemical Identifiers

753-73-1
219.68
PKKGKUDPKRTKLJ-UHFFFAOYSA-L
12955
C[Sn](C)(Cl)Cl
C2H6Cl2Sn
MFCD00000517
dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity
dichloro(dimethyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P260-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311-H314-H330-H360-H372

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 212-039-2

RTECSNumber : WH7245000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only