missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl sulfoxide, Spectrophotometric Grade, 99.9+%, Thermo Scientific Chemicals

$214.67 - $642.62

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA32434K2
View Documents Promotion Details
Thermo Scientific Chemicals
032434K2
1 L
Each for $214.67
 
AA32434M1
View Documents Promotion Details
Thermo Scientific Chemicals
032434M1
4 x 1 L
Each for $642.62
 
Description

Description

Dimethyl sulfoxide is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67-68-5
78.13
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
CS(C)=O
C2H6OS
MFCD00002089
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CHEBI:28262

Specifications

18.4°C
Colorless
1.101 g/cm3 at 20°C
1 L
Mild
1.479
C2H6OS
2.5 hPa (2mm Hg) at 20°C
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methylsulfinylmethane
679
78.13
Dimethyl sulfoxide
67-68-5
Colorless liquid
189°C
87°C (189°F)
Liquid
>99.9%
MFCD00002089
506008
Miscible with water,ethanol,ether,benzene,chloroform and acetone.
CS(C)=O
78.13
CHEBI:28262
Spectrophotometric
Safety and Handling

Safety and Handling

P210-P370+P378q-P501c

H227

EINECSNumber : 200-664-3

RTECSNumber : PV6210000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only