Learn More
Dichloromethane, Spectrophotometric Grade, 99.7+%, stab. with amylene, Thermo Scientific Chemicals
$109.18 - $315.32
Chemical Identifiers
CAS | 75-09-2 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Description
Dichloromethane is used in spectrophotometry and environmental testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
6344 | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
CHEBI:15767 | |
ClCCl |
Specifications
-95°C | |
1.325 g/mL | |
1 L | |
Liquid | |
CH2Cl2 | |
UN1593 | |
14,6063 | |
Miscible with ethyl acetate,alcohol,hexanes,methanol,diethyl ether,n-octanol,acetone benzene,carbon tetrachloride,diethyl ether and chloroform. Immiscible with water. | |
ClCCl | |
84.93 | |
CHEBI:15767 | |
≥99.7% | |
Dichloromethane, Stabilized with amylene |
75-09-2 | |
39°C to 40°C | |
Sweet | |
1.4244 | |
MFCD00000881 | |
1730800 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
6344 | |
84.93 | |
Spectrophotometric |
Safety and Handling
GHS H Statement
H351
Suspected of causing cancer.
P201-P202-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H336-H350-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DICHLOROMETHANE
EINECSNumber : 200-838-9
RTECSNumber : PA8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only