Learn More
Dichloromethane, HPLC Grade, 99.7+%, stab. with amylene, Thermo Scientific Chemicals
$62.19 - $357.93
Chemical Identifiers
CAS | 75-09-2 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA22917AK
|
Thermo Scientific Chemicals
022917AK |
250 mL |
Each for $62.19
|
|
|||||
AA22917K2
|
Thermo Scientific Chemicals
022917K2 |
1 L |
Each for $117.23
|
|
|||||
AA22917M1
|
Thermo Scientific Chemicals
022917M1 |
4 x 1 L |
Each for $357.93
|
|
|||||
AA22917K7
|
Thermo Scientific Chemicals
022917K7 |
4 L |
Each for $279.62
|
|
|||||
Description
Dichloromethane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
ClCCl |
Specifications
75-09-2 | |
Colorless liquid | |
39°C to 40°C | |
Sweet | |
1.4244 | |
MFCD00000881 | |
1730800 | |
Miscible with ethyl acetate,alcohol,hexanes,methanol,diethyl ether,n-octanol,acetone benzene,carbon tetrachloride,diethyl ether and chloroform. Immiscible with water. | |
ClCCl | |
84.93 | |
HPLC |
Colorless | |
1.325 | |
250 mL | |
Liquid | |
CH2Cl2 | |
475 hPa (356mm Hg) at 20°C | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
84.93 | |
Dichloromethane |
Safety and Handling
P201-P202-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H336-H350-H373
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 200-838-9
RTECSNumber : PA8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only