Learn More
Di-n-butyltin dichloride, 96%, Thermo Scientific Chemicals
Organotin intermediate
$44.22 - $402.15
Chemical Identifiers
CAS | 683-18-1 |
---|---|
Molecular Formula | C8H18Cl2Sn |
Molecular Weight (g/mol) | 303.842 |
MDL Number | MFCD00000518 |
InChI Key | RJGHQTVXGKYATR-UHFFFAOYSA-L |
Synonym | dibutyltin dichloride, dibutyldichlorotin, di-n-butyltin dichloride, stannane, dibutyldichloro, dibutyltin chloride, dichlorodibutyltin, dibutyl dichloro stannane, dichlorodibutylstannane, dibutyltindichloride, dibutyl tin dichloride |
PubChem CID | 12688 |
IUPAC Name | dibutyl(dichloro)stannane |
SMILES | CCCC[Sn](CCCC)(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1411609
|
Thermo Scientific Chemicals
01411609 |
10 g |
Each for $44.22
|
|
|||||
AA1411630
|
Thermo Scientific Chemicals
01411630 |
250 g |
Each for $149.48
|
|
|||||
AA14116A1
|
Thermo Scientific Chemicals
014116A1 |
1 kg |
Each for $402.15
|
|
|||||
Description
Organotin intermediate.Di-n-butyltin dichloride is used as an organotin intermediate. It serves as heat and light stabilizer for polyvinyl chloride plastics. It is also used in the preparation of methyl 2-(methoxycarbonylmethyl)-5-iodobenzene by reacting with 2,5-diiodobenzoic acid. It acts as an esterification catalyst and a veterinary vermicide and tapeworm remedy. Further, it is used as ion exchange agent, process regulators and processing aids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
683-18-1 | |
303.842 | |
RJGHQTVXGKYATR-UHFFFAOYSA-L | |
12688 | |
CCCC[Sn](CCCC)(Cl)Cl |
C8H18Cl2Sn | |
MFCD00000518 | |
dibutyltin dichloride, dibutyldichlorotin, di-n-butyltin dichloride, stannane, dibutyldichloro, dibutyltin chloride, dichlorodibutyltin, dibutyl dichloro stannane, dichlorodibutylstannane, dibutyltindichloride, dibutyl tin dichloride | |
dibutyl(dichloro)stannane |
Specifications
683-18-1 | |
White | |
135°C (10 mmHg) | |
Acrid | |
1.4991 | |
MFCD00000518 | |
UN2928 | |
dibutyltin dichloride, dibutyldichlorotin, di-n-butyltin dichloride, stannane, dibutyldichloro, dibutyltin chloride, dichlorodibutyltin, dibutyl dichloro stannane, dichlorodibutylstannane, dibutyltindichloride, dibutyl tin dichloride | |
RJGHQTVXGKYATR-UHFFFAOYSA-L | |
dibutyl(dichloro)stannane | |
12688 | |
96% | |
Di-n-butyltin dichloride |
39°C to 41°C | |
1.36 g/mL | |
168°C (334°F) | |
C8H18Cl2Sn | |
[CH3(CH2)3]2SnCl2 | |
10 g | |
Moisture sensitive | |
Insoluble in cold water; Hydrolyzed by hot water; Soluble in organic solvents;Insoluble in cold water; Soluble in benzene,toluene,ether and alcohol. | |
CCCC[Sn](CCCC)(Cl)Cl | |
303.842 | |
303.83 | |
Crystalline |
Safety and Handling
GHS H Statement
H301-H330-H360-H372-H341-H314-H312
Toxic if swallowed.
Fatal if inhaled.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
P201-P202-P260-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H312-H314-H330-H341-H360FD-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber : 211-670-0
RTECSNumber : WH7100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only