Learn More
Di-n-butyltin bis(2-ethylhexanoate), Thermo Scientific Chemicals
$64.49 - $714.25
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA7112606
|
Thermo Scientific Chemicals
07112606 |
5 g |
Each for $64.49
|
|
|||||
AA7112614
|
Thermo Scientific Chemicals
07112614 |
25 g |
Each for $216.51
|
|
|||||
AA7112622
|
Thermo Scientific Chemicals
07112622 |
100 g |
Each for $714.25
|
|
|||||
Description
Di-n-butyltin bis(2-ethylhexanoate) is used in silanol condensation reactions for caulk and sealant applications and in the production of silicones. It is also used in one or two component systems which are compatible with aromatic solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2781-10-4 | |
MFCD00015261 |
Specifications
2781-10-4 | |
1.070 g/mL | |
Odorless | |
MFCD00015261 | |
UN3146 | |
Soluble in mineral oil, acetone, xylene | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)O)N | |
363.48 | |
519.34 | |
Di-n-butyltin bis (2-ethylhexanoate) |
57°C to 59°C | |
220°C (2 mmHg) | |
C22H21NO2S | |
5 g | |
s-trityl-l-cysteine, +-s-trityl-l-cysteine, h-cys trt-oh, s-tritylcysteine, tritylcysteine, 3-tritylthio-l-alanine, 2r-2-amino-3-triphenylmethyl sulfanyl propanoic acid, l-alanine, 3-tritylthio, stlc, s-triphenylmethyl-l-cysteine | |
DLMYFMLKORXJPO-UHFFFAOYNA-N | |
(2R)-2-amino-3-tritylsulfanylpropanoic acid | |
76044 | |
Lumps |
Safety and Handling
GHS H Statement
H301-H310-H361
Toxic if swallowed.
Fatal in contact with skin.
Suspected of damaging fertility or the unborn child.
P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
H301-H340-H360-H372-H500
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 220-481-2
RTECSNumber : WH6714500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only