missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Decon™ Proguard™ Professional Hand Cream
Specially formulated lotion moisturizes dry, cracked hands and guards against latex glove irritation
$22.40 - $87.73
Specifications
Product Type | Professional Hand Cream |
---|---|
Color | White |
Form | Cream |
For Use With (Application) | Specially formulated lotion moisturizes dry, cracked hands and guards against latex glove irritation |
Quantity | 1 Ea. |
Description
- Does not contain grease, oil, perfume, aloe, or other ingredients that can compromise the integrity of latex gloves
- Absorbs instantly, leaving skin soft
Specifications
Professional Hand Cream | |
Cream | |
1 Ea. |
White | |
Specially formulated lotion moisturizes dry, cracked hands and guards against latex glove irritation |