missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
α-D-Methylglucoside, 99%, Thermo Scientific Chemicals
$62.43 - $169.88
Chemical Identifiers
CAS | 97-30-3 |
---|---|
Molecular Formula | C7H14O6 |
Molecular Weight (g/mol) | 194.18 |
MDL Number | MFCD00064086 |
InChI Key | HOVAGTYPODGVJG-IECVIRLLSA-N |
Synonym | methyl alpha-d-glucopyranoside, methyl alpha-d-glucoside, alpha-methylglucoside, alpha-methyl-d-glucoside, alpha-methyl d-glucose ether, methyl-alpha-d-glucopyranoside, alpha-d-methylglucoside, me alpha-glc, alpha-d-methyl glucoside, unii-qcf122nf3r |
PubChem CID | 7568545 |
IUPAC Name | (2R,3R,4R,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
SMILES | COC1C(C(C(C(O1)CO)O)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC173841000
|
Thermo Scientific Chemicals
173841000 |
100 g | Plastic bottle |
Each for $62.43
|
|
||||
AC173845000
|
Thermo Scientific Chemicals
173845000 |
500 g | Plastic bottle |
Each for $169.88
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
97-30-3 | |
194.18 | |
HOVAGTYPODGVJG-IECVIRLLSA-N | |
7568545 | |
COC1C(C(C(C(O1)CO)O)O)O |
C7H14O6 | |
MFCD00064086 | |
methyl alpha-d-glucopyranoside, methyl alpha-d-glucoside, alpha-methylglucoside, alpha-methyl-d-glucoside, alpha-methyl d-glucose ether, methyl-alpha-d-glucopyranoside, alpha-d-methylglucoside, me alpha-glc, alpha-d-methyl glucoside, unii-qcf122nf3r | |
(2R,3R,4R,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
Specifications
97-30-3 | |
100.0 | |
White | |
0.1% max. (60 to 70°C) | |
98.5% min. (ELSD) (HPLC) | |
C7H14O6 | |
100 g | |
15, 6152 | |
+ 158.50 | |
Solubility in water: 108g/100mL (20 c). Other solubilities: practically insoluble in ether,5.2g/100g methanol (20 c),7.3g/100g alcohol 80% (17 c) | |
COC1C(C(C(C(O1)CO)O)O)O | |
194.18 | |
194.18 | |
Fine Crystalline Powder |
98.5 | |
169.0°C to 171.0°C | |
200.0°C (0.2 mmHg) | |
Authentic | |
Plastic bottle | |
MFCD00064086 | |
31, 179 | |
+158.50° (20°C c=10,water) | |
methyl alpha-d-glucopyranoside, methyl alpha-d-glucoside, alpha-methylglucoside, alpha-methyl-d-glucoside, alpha-methyl d-glucose ether, methyl-alpha-d-glucopyranoside, alpha-d-methylglucoside, me alpha-glc, alpha-d-methyl glucoside, unii-qcf122nf3r | |
HOVAGTYPODGVJG-IECVIRLLSA-N | |
(2R,3R,4R,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol | |
7568545 | |
99% | |
α-D-Methylglucoside, 99% |
Safety and Handling
EINECSNumber : 202-571-3
TSCA : TSCA
RUO – Research Use Only