Thermo Scientific™ Cytospin™ 4 Cytocentrifuge Accessories
Use these accessories with the Thermo Scientific™ Cytospin™ 4 Cytocentrifuge.
$74.35 - $1,430.00
Specifications
For Use With (Equipment) Cytospin™ 4 Cytocentrifuge
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
SH59910019 View Documents Thermo Scientific™
59910019
Sealed Head Seals Pack of 3 Each for $74.35
SH59910018 View Documents Thermo Scientific™
59910018
Sealed Head Rotor Each for $1,430.00
Description
Description
Use these accessories with the Thermo Scientific™ Cytospin™ 4 Cytocentrifuge.
  • Shorten preparation time by having a second sealed head ready when the first finishes processing
  • Autoclavable
  • 12-chamber head seals completely
Specifications
Specifications
Cytospin™ 4 Cytocentrifuge