missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals H58852AD

Catalog No. AAH58852AD



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary intermediate.

Solubility
Reacts with water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
Specifications

Specifications

Cyclohexylzinc bromide
7565-57-3
C6H11BrZn
50 mL
Air sensitive
Reacts with water.
C1CC[CH-]CC1.[Zn+]Br
228.438
228.44
Packaged under Argon in resealable ChemSeal™ bottles
0.963
MFCD00671993
UN3399
cyclohexylzinc bromide, cyclohexylzinc bromide solution, bromozinc 1+ ,cyclohexane, bromo cyclohexyl zinc, cyclohexylzinc bromide, 0.50 m in thf, cyclohexylzinc bromide solution, 0.5 m in thf, cyclohexylzinc bromide 0.5 m in tetrahydrofuran, cyclohexylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
PVURAUIMVICLOH-UHFFFAOYSA-M
bromozinc(1+);cyclohexane
4385789

Chemical Identifiers

7565-57-3
228.438
PVURAUIMVICLOH-UHFFFAOYSA-M
4385789
C1CC[CH-]CC1.[Zn+]Br
C6H11BrZn
MFCD00671993
cyclohexylzinc bromide, cyclohexylzinc bromide solution, bromozinc 1+ ,cyclohexane, bromo cyclohexyl zinc, cyclohexylzinc bromide, 0.50 m in thf, cyclohexylzinc bromide solution, 0.5 m in thf, cyclohexylzinc bromide 0.5 m in tetrahydrofuran, cyclohexylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
bromozinc(1+);cyclohexane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302-H314-H335-H336-H351

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only