missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Transwell™ Multiple Well Plate with Permeable Polycarbonate Membrane Inserts
Patented self-centering design provides independent access to both apical and basolateral membranes of the cell monolayer. Corning™ Transwell™Multiple Well Plate with Permeable Polycarbonate Membrane Inserts provide an ideal solution for studying anchorage-dependent and -independent cells.
Supplier: Corning™ C3422
Description
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
You can use cell culture inserts to:
- Produce a cell culture environment that closely resembles an in vivo state
- Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
- Co-culture cells with or without cell-to-cell contact
- Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 μm thick translucent membrane
- Pore sizes ranging from 0.4 μm to 8 μm diameters
- Treated for optimal cell attachment
- Supplied in multiple well plates
- Membrane must be stained for cell visibility
- Sterile
Specifications
12 inserts/24 well plate | |
Polycarbonate | |
8 μm | |
Epithelial and endothelial cell culture; transport, migration/invasion, co-culture, and microbial pathogenesis studies | |
12/Pk. | |
TC-Treated | |
TC-Treated | |
24 |
0.33 cm2 | |
0.6 mL | |
Pre-loaded Microporous Insert in Tissue Culture Plate | |
6.5 mm | |
48/Cs. | |
Clear | |
Sterile |