Corning™ Costar™ Sterile Disposable Reagent Reservoirs
Manufactured from modified polystyrene for strength. Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipetters.
$469.79 - $676.83
Specifications
Material Polystyrene
Sterility Sterile
Length (Metric) 145 mm
Disposable Single-use
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Packaging Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Packaging Price Quantity  
07-200-130
View Documents Promotion Details
Corning™
C4873
100 mL Individually Wrapped
Case of 100 Each for $676.83
07-200-127 Edge
View Documents Promotion Details
Corning™
C4870
50 mL Bulk
Case of 200 Each for $469.79
Edge
07-200-128
View Documents Promotion Details
Corning™
C4871
50 mL Individually Wrapped
Case of 100 Each for $635.09
07-200-129
View Documents Promotion Details
Corning™
C4872
100 mL Bulk
Case of 200 Each for $532.23
Description
Description
Manufactured from modified polystyrene for strength. Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipetters.
Costar™ reagent reservoirs are ideal for repetitively filling multichannel pipettors.
  • Manufactured from modified polystyrene (white)
  • Individually wrapped, sterile
  • Disposable
Specifications
Specifications
Polystyrene
Sterile
145 mm
Single-use
SDS
missing translation for 'documents'
missing translation for 'documents'