PYREX™ VISTA™ Rimless Culture Tubes
Offer convenient plugging and rack storage.
$24.00 - $75.30
Specifications
Design Type Fire Polished
Closure Type Without
Material Borosilicate Glass
Sterility Non-sterile
Disposable No
View More Specs
Products 8
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Label Type Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Label Type Price Quantity  
S76111C View Documents Fisher Scientific
7082013EM
9mL 13mm 100mm Marking Spot Pack of 50 Each for $27.35
Case of 4 Pack for $109.35
S76111E View Documents Fisher Scientific
70820-16X/EMD
15mL 16mm 125mm Marking Spot Pack of 50 Each for $38.50
Case of 4 Pack for $153.90
S76111D View Documents Fisher Scientific
7082016EMD
11mL 16mm 100mm Marking Spot Pack of 50 Each for $34.85
Case of 4 Pack for $139.50
S76111F View Documents Fisher Scientific
7082016XX
20mL 16mm 150mm Marking Spot Pack of 50 Each for $41.20
Case of 4 Pack for $164.70
S76111G View Documents Fisher Scientific
7082018EM
27mL 18mm 150mm Marking Spot Pack of 50 Each for $42.90
Case of 4 Pack for $171.45
S76111J View Documents Fisher Scientific
7082022EM
50mL 22mm 175mm None Pack of 50 Each for $75.30
Case of 4 Pack for $301.05
S76111M View Documents Fisher Scientific
708206EMD
0.5mL 6mm 50mm Marking Spot Pack of 50 Each for $24.00
Case of 4 Pack for $95.85
S76111H View Documents Fisher Scientific
7082020EMD
34mL 20mm 150mm Marking Spot Pack of 50 Each for $49.95
Case of 4 Pack for $199.80
Description
Description
  • Reusable tubes
  • Fire-polished ends
  • Blue marking spot (except 0.86 in. [22mm] and 0.98 in. [25mm] O.D. tubes)
Specifications
Specifications
Fire Polished
Borosilicate Glass
No
No
Round
Without
Non-sterile
For convenient plugging and rack storage
No
Untreated