missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Premium Fetal Bovine Serum
FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
Supplier: Corning™ 35016CV
Description
- Sera is aseptically collected and processed in a cGMP facility
- Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
- Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
- Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
- Premium FBS is sourced from U.S. herds
Specifications
cGMP | |
-25° to -15°C | |
500 mL | |
Triple-filtered, 0.1 μm | |
Premium | |
Used as a supplement to cell culture media to help sustain cell growth and metabolism. This product is for research use only. | |
<30 mg/dL | |
7.8 ±0.8 | |
USDA Inspected |
Caramel/Amber | |
Fetal Bovine Serum | |
Sterile | |
Bovine Fetus | |
≤20EU/ mL | |
Yes | |
315 ± 35mOsm/kg H20 | |
30 to 50g/L |