missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Polyethylene Terephthalate (PET) Centrifuge Tubes

Manufactured from crystal clear rigid PET for optimum visibility. Corning™ Polyethylene Terephthalate (PET) Centrifuge Tubes have conical bottoms and high-density polyethylene plug seal caps providing a premium centrifuge tube.

$508.51 - $854.53

Specifications

Color Clear
Disposable Yes
Material PET
Sterility Sterile
Label Type Large White Marking Area
View More Specs

Products 3
Catalog Number Mfr. No. Capacity (Metric) Graduations Diameter (Metric) Outside Length (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Graduations Diameter (Metric) Outside Length (Metric) Price Quantity  
05-538-51A Accessories View Documents Corning™
C430053
15mL 1.5 to 14.5mL 17mm 119mm Case of 500 Each for $508.51
05-538-49 Accessories View Documents Corning™
C430304
50mL Yes 30mm 114mm Case of 500 Each for $854.53
05-538-51 Accessories View Documents Corning™
C430055
15mL Yes 17mm 19mm Case of 500 Each for $533.06
Description

Description

  • Conical bottom
  • Clear polyethylene tetraphthalate (PET)
  • Sterile, certified RNase-/DNase-free and nonpyrogenic
  • Wide neck with threaded top
  • Max RCF: 3600 x G
  • Graduated (5 to 50mL) at 2.5mL intervals
  • 29.1mm O.D. (approx.)
  • 115.5mm L (approx.)
  • Disposable
  • High-density polyethylene plug seal cap (approx. 35.2mm; orange)

Order Info

Rack included

Specifications

Specifications

Clear
PET
Large White Marking Area
3,600 x g
Yes
Sterile
Conical