missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Laminin/Entactin High Concentration Extracellular Matrix Protein

Provides a highly defined 3D culture environment for cell differentiation assays

Manufacturer:  Corning™ C354259

Catalog No. 08-774-555


Alert:

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.


Description

Description

  • Source: Engelbreth-Holm-Swarm (EHS) mouse tumor
  • Formulation: Frozen in Dulbecco's Phosphate-Buffered Saline
  • Purity >90% by SDS-PAGE
  • Tested for ability to initiate neurite outgrowth of NG-108 Mouse neuroblastoma cells.
  • Membrane filtered (0.2mm) preparation
  • Molecular Weight: 900kDa (unreduced)

Specifications

Specifications

Bioassay, ELISA Standard
High concentration Laminin/Entactin complex frozen in Dulbecco's PBS
SDS-PAGE
10.5mg
-70°C
Liquid
Laminin/Entactin
>95% by SDS-PAGE
Engelbreth Holm Swarm (EHS) Mouse tumor
Mouse
Documents

For research use only.