missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ HYPERFlask™ M Cell Culture Vessels

Designed specifically for manual use, high-yield and high-performance. Specifically designed for manual use. Corning™ HYPERFlask™ M Cell Culture Vessel uses a multi-layer gas permeable growing surface for efficient gas exchange.

$1,405.38 - $8,786.56

Specifications

Capacity (Metric) 560mL
Closure Color Orange
Closure Material HDPE
Closure Type Flat
Culture Area 1720cm2
View More Specs

Products 4
Catalog Number Mfr. No. Surface Coating No. per Pack No. per Case Price Quantity  
Catalog Number Mfr. No. Surface Coating No. per Pack No. per Case Price Quantity  
09-761-23 View Documents Corning™
C10034
CellBIND™ treated 4.0 24 Case of 24 Each for $8,786.56
09-761-22 View Documents Corning™
C10020
CellBIND™ treated 4 4 Case of 4 Each for $1,464.38
13-700-420 View Documents Corning™
C10030
CellBIND™ treated 1.0 4 Case of 4 Each for $1,464.38
13-700-421 View Documents Corning™
C10031
Non-Treated 1 4 Case of 4 Each for $1,405.38
Description

Description

The novel Corning HYPERFlask cell culture vessel offers 1720cm2 growth area in the footprint of a traditional 175cm2 flask. 

 

  • Adapter Grid: design allows for faster filling and emptying while reducing foam generation
  • Serial Number: each individual flask is traceable by a serial number that can be read by the unaided eye or by a handheld barcode reader
  • Innovative 10 Layer Design: 10 interconnected polystyrene growth surfaces
  • Optimal Growth: Corning CELLBIND™ surface-treated gas permeable polystyrene for superior cell attachment and growth
  • Increased Cell Yield: 10-fold higher cell yield increases productivity and capacity
  • Time and Space Savings: Reduce processing time and incubator storage space by handling one flask compared to 10 traditional 175cm2 flasks
  • Made with Class VI Materials: molded and assembled in a Certified Class 100,000 cleanroom

 

Order Info

Supplied double bagged.

Specifications

Specifications

560mL
HDPE
1720cm2
Straight
No
Sterile
Orange
Flat
PS (Polystyrene)
560mL
Rectangular