Corning™ Magnetic Stir Bar Retriever
Designed for safe removal of heated stir bars in liquids on hot plate stirrers. Corning™ Magnetic Stir Bar Retriever is effective with all stirring models.
Manufacturer:  Corning™ C6970SR
Catalog No. 14-972-50
Description
Description
  • High powered magnet removes stir bars from glass and plastic containers
  • Ergonomic design for easy retrieval
Note: Remove container from magnetic stirrer before using retriever; the stirrer magnet cancels out the power in the retriever magnet.
Specifications
Specifications
All stirring models
Magnetic Stir Bar Retriever
Documents