missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) sulfate pentahydrate, 98+%, ACS reagent, ACROS Organics™

$130.10 - $532.03

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.677
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper;sulfate;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=O)[O-].[Cu+2]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC423615000 SDS View Documents Acros Organics
423615000
500g Plastic bottle Each for $130.10
AC423610030 SDS View Documents Acros Organics
423610030
3kg Plastic bottle Each for $532.03
AC423610100 SDS View Documents Acros Organics
AC423610100
10kg Plastic Bucket N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

7758-99-8
249.677
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
CHEBI:31440
O.O.O.O.O.[O-]S(=O)(=O)[O-].[Cu+2]
CuH10O9S
JZCCFEFSEZPSOG-UHFFFAOYSA-L
24463
copper;sulfate;pentahydrate

Specifications

7758-99-8
CuH10O9S
01,164; 02,89; 05,162; 06,141; 14,267
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper;sulfate;pentahydrate
24463
249.68
≥98%
50ppm max.
0.005% max.
50ppm max.
Plastic bottle
200ppm max.
Blue
500g
98+%
CuSO4·5H2O
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
O.O.O.O.O.[O-]S(=O)(=O)[O-].[Cu+2]
249.677
CHEBI:31440
Fine Crystals
ACS Reagent
10ppm max.
30ppm max.
0.002% max.
100ppm max.
Solubility in water: 320g/L (20°C). Other solubilities: soluble in methanol, slightly soluble in alcohol and glycerol, insoluble in ethanol
110.0°C
Copper(II) sulfate pentahydrate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning