Learn More
Copper bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
$72.55 - $809.60
Chemical Identifiers
CAS | 291300-50-0 |
---|---|
Molecular Formula | C2CuF6NO4S2 |
Molecular Weight (g/mol) | 343.681 |
MDL Number | MFCD23380170 |
InChI Key | MSFHCZUIBARZOS-UHFFFAOYSA-N |
Synonym | copper trifluoromethanesulfonimide |
PubChem CID | 134159300 |
IUPAC Name | bis(trifluoromethylsulfonyl)azanide;copper(1+) |
SMILES | C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Cu+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA47162MD
|
Thermo Scientific Chemicals
047162MD |
250 mg |
Each for $72.55
|
|
|||||
AA4716203
|
Thermo Scientific Chemicals
04716203 |
1 g |
Each for $228.48
|
|
|||||
AA4716206
|
Thermo Scientific Chemicals
04716206 |
5 g |
Each for $809.60
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
291300-50-0 | |
343.681 | |
MSFHCZUIBARZOS-UHFFFAOYSA-N | |
134159300 | |
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Cu+] |
C2CuF6NO4S2 | |
MFCD23380170 | |
copper trifluoromethanesulfonimide | |
bis(trifluoromethylsulfonyl)azanide;copper(1+) |
Specifications
291300-50-0 | |
C2CuF6NO4S2 | |
MFCD23380170 | |
UN3261 | |
copper trifluoromethanesulfonimide | |
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Cu+] | |
343.681 | |
343.68 | |
Copper bis(trifluoromethylsulfonyl)imide |
Blue | |
[(CF3SO2)2N]Cu | |
250 mg | |
Very Hygroscopic | |
MSFHCZUIBARZOS-UHFFFAOYSA-N | |
bis(trifluoromethylsulfonyl)azanide;copper(1+) | |
134159300 | |
Liquid or Solid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only