Copan Diagnostics CRYOBANK™Bead System
Easy-to-use system for freezing bacterial cultures for later retrieval
Manufacturer:  Copan Diagnostics CRYOM
 View more versions of this product
Catalog No. 14-906-51
Description
Description
  • Cryogenic medium consists of a phosphate buffered saline supplemented with sucrose, glycerol, and peptone
  • Can be stored for long periods at -60° to -80°C
  • Easy to store in unique storage boxes designed to fit freezer racks
  • Comes with 64 vials
  • Easy to locate
  • Convenient, versatile and reliable
Specifications
Specifications
Assorted Colors
For freezing bacterial cultures for later retrieval
64/Pk.
Culture Tube
Documents