Fisherbrand™ Graduated Glass Bottle Beakers with White Polypropylene PE Foam/PTFE Cap
Environmentally sensitive bottles help eliminate waste. Fisherbrand™ Clear Graduated Clear Graduated Boston Round Bottles with White Polypropylene PE Foam/PTFE Lined Cap ensures product integrity for long term storage.
$87.40 - $154.60
Specifications
Liner Material PE Foam/PTFE
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Clear, Clear
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Closure Size Quantity Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Quantity Price Quantity  
02-911-458A View Documents Fisherbrand™
ALP263467
30mL 48/Cs. Case of 48 Each for $87.40
02-911-459A View Documents Fisherbrand™
ALP263475
60mL 48/Cs. Case of 48 Each for $100.27
02-911-462A View Documents Fisherbrand™
ALP263459
480mL 24/Cs. Case of 24 Each for $154.60
02-911-461A View Documents Fisherbrand™
ALP263487
240mL 24/Cs. Case of 24 Each for $101.63
Description
Description
Graduated Medium Rounds are excellent multipurpose containers for use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents. These clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling. The wide mouth facilitates easy mixing, handling, and transfer of contents. Graduated in both milliliters and ounces.
Clear Graduated Boston Round Bottles
  • Feature a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Clear (flint) glass is appropriate when visibility is important but protection from UV rays is not necessary
 Polypropylene PE Foam/PTFE Lined Caps
  • Offer good impact strength, thermal resistance and pliability
  • Autoclavable
  • Liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • Liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference
Specifications
Specifications
PE Foam/PTFE
Screw
Clear, Clear
Wide
Bottle, Graduated, Graduated Medium Round Bottle
Polypropylene
White
Glass, Glass
Round