Thermo Scientific™ CL-XPosure™ Film (X-Ray Film)
Capture results of ECL-based Western blots and other chemiluminescent protein or nucleic acid assays using this economically priced, clear-blue X-ray film.
$256.00 - $708.00
Specifications
Formulation Blue-backed, silver-emulsion, two-sided autoradiography film
View More Specs
Products 5
Catalog Number Mfr. No. Description Quantity Price Quantity  
Catalog Number Mfr. No. Description Quantity Price Quantity  
PI34089 View Documents Thermo Scientific™
34089
CL-XPosure Film, 7 x 9.5in. (18 x 24cm) 100 sheets Each for $324.00
PI34090 View Documents Thermo Scientific™
PI34090
CL-XPosure Film, 5 x 7in. (13 x 18cm) 100 sheets Each for $256.00
PI34091 View Documents Thermo Scientific™
PI34091
CL-XPosure Film, 8 x 10in. (20 x 25cm) 100 sheets Each for $419.00
PI34097 View Documents Thermo Scientific™
34097
CL-XPosure Film, 9.5 x 11.8in. (24 x 30cm) 100 sheets Each for $367.00
PI34099 View Documents Thermo Scientific™
34099
CL-XPosure Film, 14 x 17in. (35 x 40cm) 100 sheets Pack of 100 Each for $708.00
Description
Description
Thermo Scientific™ CL-XPosure Film is an economically priced clear-blue X-ray film especially for detection and documentation of ECL-based Western blots and other chemiluminescent protein or nucleic acid assay methods.
CL-XPosure Film is an excellent phototgraphic film for use with enhanced chemiluminescence (ECL) substrates for horseradish peroxidase (HRP) or alkaline phosphatase (AP). Convenient sizes are offered for protein Western blotting, standard film cassettes and film developing equipment. The autoradiography film also works well for chemiluminescent or isotopic DNA and RNA methods, including Southern and Northern blotting and gel-shift assays (EMSA). The quality and price of this film make it the smart choice for life science research applications.
Highlights:
  • High performance – just as sensitive as other commercially available autoradiography films
  • Affordable – costs up to 75% less than other brands of comparable quality X-ray film
  • Convenient – available in five sheet sizes in packages containing 50 or 100 non-interleaved sheets
  • Perfect match – validated for use with Thermo Scientific SuperSignal Chemiluminescent Substrates and Thermo Scientific Pierce ECL Chemiluminescent Substrate
Ordering Information:
Available in 5 × 7in., 7 × 9.5in. , 9.5 × 11in., or 14 × 17in. sheets
Specifications
Specifications
Blue-backed, silver-emulsion, two-sided autoradiography film