Cincinnati Surgical Stainless Steel Fitment Handle, Size 3
Made from British stainless steel
Supplier: Cincinnati Surgical 073S
Description
- For use with blades 9 to 16
Specifications
Stainless Steel | |
Scalpel Handle | |
# 3 |