Cincinnati Surgical Multi Use Blade Remover
Assists in the safe removal of blades and disposal of contents
Supplier:  Cincinnati Surgical 48
Catalog No. 19200260
Description
Description
  • Greatly reduces risk of injury and infection
  • Contents may be safely autoclaved and disposed
  • Single size
  • Plastic composition
Specifications
Specifications
Plastic
Blade Remover