Chromatotank™ TLC Developing Tank with Latch Lid
Clear glass tank provides complete visibility of contents at all times
Manufacturer:  Gen Glassblowing Inc GCB609
Catalog No. 05-719-28C
Description
Description
  • Heavyweight construction and ground flat bottom lend stability against accidental tipping and withstand the rigors of use
  • Top lip of tank is also ground flat and beveled inside and outside to eliminate any sharp edges
  • Matching ground-glass lid fits firmly on tank and is held in place by stainless-steel latching mechanism
  • Lid has handle for easy removal
  • Six-plate aluminum rack fits snugly in tank and holds plates steady during developing
  • Interior dimensions: 3 3/8L × 4 3/4W × 4 1/2 in.D (8.6 × 12 × 11.5cm); weight of tank with lid: 15 lb. (6.8kg)
Documents