missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform/isoamyl alcohol 24:1, for molecular biology, DNAse, RNAse and Protease fr, ACROS Organics™

Manufacturer:  Acros Organics 327155000

Catalog No. AC327155000



Description

Description

For molecular biology
Specifications

Specifications

Chloroform/isoamyl alcohol 24:1(v/v)
67-66-3,67-66-3,123-51-3
MFCD00171265
Solubility in water: 8g/L (20°C).
C(Cl)(Cl)Cl
119.369
CHEBI:35255
Chloroform component:%gt; 99.9%, isoamyl alcohol component: 99.0% min. (by GC)
1.4530g/mL
Yes
Solubility in water: 8g/L (20°C).
0.56 mPa.s (20°C)
Undesignated
500mL
DNase, RNase and Protease Free
CHCl3
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
Solution
Molecular Biology
none detected
Glass bottle
1.453
62.0°C
-63.0°C

Chemical Identifiers

67-66-3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
chloroform
CHCl3
MFCD00171265
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
C(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation.

GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray. Use personal protective equipment as required. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

SDS Documents