missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform, HPLC Grade, >99.5% min, Alfa Aesar™

$63.58 - $240.84

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.369
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name chloroform
SMILES C(Cl)(Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA22920K2 SDS View Documents Alfa Aesar
22920K2
1L Each for $63.58
AA22920K7 SDS View Documents Alfa Aesar
22920K7
4L Each for $182.50
AA22920M1 SDS View Documents Alfa Aesar
22920M1
4 x 1L Each for $240.84
Specifications

Chemical Identifiers

67-66-3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
chloroform
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
C(Cl)(Cl)Cl

Specifications

67-66-3
>99.5%
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
119.38
HPLC
Characteristic
Colorless
Chloroform
Colorless liquid
CHCl3
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
C(Cl)(Cl)Cl
119.369
CHEBI:35255
Liquid
1.492g/cm3 at 20°C
210 hPa (158mm Hg) at 20°C
1L
Safety and Handling

Safety and Handling

H351-H373-H302-H315: Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Causes skin irritation.

DOTInformation : DOT Class: 6.1; Packing Group: III

EINECSNumber : 200-663-8