missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform (Approx. 0.75% Ethanol as Preservative/Technical), Fisher Chemical

$616.50 - $2,127.36

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.369
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name chloroform
SMILES C(Cl)(Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
C295-20 SDS View Documents Thermo Scientific
BPC29520
20L Steel Pail Each for $2,127.36
C295-4 SDS Accessories View Documents Thermo Scientific
BPC2954
4L Glass Bottle Each for $616.50
Case of 4 Each for $2,466.00
C295S-4 SDS View Documents Thermo Scientific
BPC295S4
4L Aluminum Can Each for $712.36
Case of 4 Each for $2,849.44
Specifications

Chemical Identifiers

67-66-3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
chloroform
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
C(Cl)(Cl)Cl

Specifications

67-66-3,64-17-5
CHCl3
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
119.38
Technical
Pass Test
213mbar at 20°C
0.56 mPa/s at 20°C
61°C
-63°C
Chloroform, Approx. 0.75% ethanol as preservative
Vapor Pressure: 213mbar at 20°C
MFCD00000826
Approx. 0.75% Ethanol as Preservative
C(Cl)(Cl)Cl
119.369
CHEBI:35255
Liquid
1.480g/cm³
Steel Pail
4.12g/mol
290°C
Colorless
20L
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. May be harmful if inhaled. May cause irritation of respiratory tract. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:2
Flammability:1
Instability:0

DOTInformation : DOT Class 6.1, : Poison