missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform (Amylene as Preservative, Pesticide), Fisher Chemical

Manufacturer:  Thermo Scientific FLC6034

Catalog No. C603-4




Specifications

Specifications

Chloroform
67-66-3,513-35-9
CHCl3
1888
With Amylene as Preservative
C(Cl)(Cl)Cl
119.369
CHEBI:35255
Liquid
1.480g/cm³
Amber Glass
Pass Test
0.56 mPa/s at 20°C
Colorless
4L
With 150-200ppm amylene as preservative
Vapor Pressure: 213mbar at 20°C
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
119.38
Pesticide
Pass Test
150 to 200ppm
213mbar at 20°C
61°C
-63°C

Chemical Identifiers

67-66-3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
chloroform
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
C(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin. Symptoms of overexposure are dizziness, headache, tiredness, nausea, unconsciousness, cessation of breathing.
<special id

Signal Word:WARNING!

SDS Documents
Accessories