missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform, 99.8%, for analysis, stabilized with amylene, ACROS Organics™

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.369
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
PubChem CID 6212
ChEBI CHEBI:35255
View More Specs

Products 2 Description

Description

For analysis
Specifications

Chemical Identifiers

67-66-3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
CHEBI:35255
CHCl3
MFCD00000826
6212

Specifications

67-66-3,513-35-9
99.8%
CHCl3
15, 2142
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
119.369
CHEBI:35255
119.38
0.02ppm max.
< 0.005%
0.1ppm max.
0.05ppm max.
5ppm max.
Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in acetone.
1.48
5 APHA max.
Tetrachloroethylene< 0.005%
0.05ppm max.
0.5ppm max.
0.56 mPa.s (20°C)
Undesignated
25L
99.8%
0.0005 meq/g max.
CHCl3
MFCD00000826
Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in acetone.
C(Cl)(Cl)Cl
0.1ppm max.
6212
< 0.005%
Liquid
1.4800g/mL
5ppm max.
0.02ppm max.
0.1ppm max.
0.05ppm max.
0.2ppm max.
approx. 100ppm amylene
for use with dithizone: Passes Test
0.1ppm max.
trichloroethylene < 0.005%
0.02% max. (K.F.)
61.0°C
-63.0°C
Chloroform, Stabilized with amylene
For analysis
Safety and Handling

Safety and Handling

GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Toxic if inhaled. Suspected of causing cancer. May cause drowsiness or dizziness. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning