Learn More
Chlorobenzene, ACS, 99.5%, Thermo Scientific Chemicals
$91.21 - $427.73
Chemical Identifiers
CAS | 108-90-7 |
---|---|
Molecular Formula | C6H5Cl |
Molecular Weight (g/mol) | 112.556 |
MDL Number | MFCD00000530 |
InChI Key | MVPPADPHJFYWMZ-UHFFFAOYSA-N |
Synonym | monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene |
PubChem CID | 7964 |
ChEBI | CHEBI:28097 |
IUPAC Name | chlorobenzene |
SMILES | C1=CC=C(C=C1)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA36401AP
|
Thermo Scientific Chemicals
036401AP |
500 mL |
Each for $91.21
|
|
|||||
AA36401K2
|
Thermo Scientific Chemicals
036401K2 |
1 L |
Each for $136.12
|
|
|||||
AA36401M1
|
Thermo Scientific Chemicals
036401M1 |
4 x 1 L |
Each for $427.73
|
|
|||||
AA36401K7
|
Thermo Scientific Chemicals
036401K7 |
4 L |
Each for $354.48
|
|
|||||
Description
Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-90-7 | |
112.556 | |
MVPPADPHJFYWMZ-UHFFFAOYSA-N | |
7964 | |
chlorobenzene |
C6H5Cl | |
MFCD00000530 | |
monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene | |
CHEBI:28097 | |
C1=CC=C(C=C1)Cl |
Specifications
-45°C | |
Colorless | |
132°C | |
28°C (82°F) | |
Liquid | |
C6H5Cl | |
UN1134 | |
14,2121 | |
Miscible with methanol and diethyl ether. Immiscible with water. | |
C1=CC=C(C=C1)Cl | |
112.556 | |
CHEBI:28097 | |
99.5% | |
Chlorobenzene |
108-90-7 | |
1.106 g/mL | |
500 mL | |
Aromatic | |
1.524 | |
MFCD00000530 | |
605632 | |
monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene | |
MVPPADPHJFYWMZ-UHFFFAOYSA-N | |
chlorobenzene | |
7964 | |
112.56 | |
ACS |
Safety and Handling
GHS H Statement
H226-H332
Flammable liquid and vapor.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
H226-H315-H332
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZENE
EINECSNumber : 203-628-5
RTECSNumber : CZ0175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only