missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium Chloride, 99.99%, (Optical Grade I), MP Biomedicals

Solutions are clear and colorless suitable for density gradient work, nucleic acid and protein purification

$110.17 - $1,875.35

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.355
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN16004125 View Documents MP Biochemicals LLC
16004125
25g Each for $110.17
 
ICN16004180 View Documents MP Biochemicals LLC
16004180
100g Each for $315.97
 
ICN16004190 View Documents MP Biochemicals LLC
16004190
500g Each for $1,192.38
 
ICN16004191 View Documents MP Biochemicals LLC
16004191
1kg Each for $1,875.35
 
Specifications

Chemical Identifiers

7647-17-8
168.355
caesium 1+ ion chloride
CHEBI:63039
[Cl-].[Cs+]
ClCs
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
cesium;chloride

Specifications

7647-17-8
ClCs
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.4
25g
99.99%
caesium 1+ ion chloride
[Cl-].[Cs+]
168.355
CHEBI:63039
Optical I
Cesium Chloride
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature